Learn More
Influenza A Virus H1N1 Hemagglutinin Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11396-100
This item is not returnable.
View return policy
Description
Hemagglutinin/HA Peptide Monoclonal antibody specifically detects Hemagglutinin/HA Peptide in Influenza virusN1 samples. It is validated for Western Blot, Immunocytochemistry
Specifications
| Hemagglutinin/HA Peptide | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 2908B | |
| 1 μg/mL (Western Blot), 3 to 15 μg/mL (Immunocytochemistry) | |
| YP_009118626 | |
| Human embryonic kidney cell, HEK293-derived influenza a virus H1N1 Hemagglutinin with a C-terminal 6-His tag, Met1-Gln529, Accession # YP_009118626 | |
| 100 μg | |
| Primary | |
| Influenza virus N1 | |
| Purified |