missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 312.00 - € 715.00
Specifications
| Antigen | ING2 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600068
|
Novus Biologicals
NBP3-05515-100ul |
100 μg |
€ 715.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18697578
|
Novus Biologicals
NBP3-05515-25ul |
25 μg |
€ 312.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ING2 Polyclonal antibody specifically detects ING2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| ING2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, DNA Repair | |
| PBS, pH 7.2, containing 40% glycerol | |
| 3622 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ING1Linhibitor of growth family, member 1-like, ING1Lp, Inhibitor of growth 1-like protein, inhibitor of growth family, member 2, inhibitor of growth protein 2, p33ING2p32 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MLGQQQQQLYSSAALLTGERSRLLTCYVQDYLECVESLPHDMQRNVSVLRELDNKYQETLKEIDDVYEKYKKEDDLNQKKRLQQLLQRAL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title