missing translation for 'onlineSavingsMsg'
Learn More

INO80E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-98592-100ul

 View more versions of this product

Product Code. 18655842

  • € 326.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

INO80E Polyclonal antibody specifically detects INO80E in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

INO80E
Polyclonal
Immunocytochemistry/ Immunofluorescence 1:100-1:500
CCDC95, coiled-coil domain containing 95, Coiled-coil domain-containing protein 95, FLJ00079, FLJ90652, INO80 complex subunit E
Produced in rabbits immunized with E. coli-derived Human INO80E fragment.
100 μg
Primary
Human
Purified
Immunofluorescence
Unconjugated
PBS (pH 7.0)
Rabbit
Antigen and protein A Affinity-purified
RUO
283899
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.