missing translation for 'onlineSavingsMsg'
Learn More

INO80E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 216.00 - € 307.65

Specifications

Antigen INO80E
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18655842
View Documents
Novus Biologicals
NBP2-98592-100ul
100 μg
ONLINE EXCLUSIVE PRICE
€ 326.00
€ 307.65 / 100µL
Save € 18.35
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18653575
View Documents
Novus Biologicals
NBP2-98592-50ul
50 μg
€ 216.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

INO80E Polyclonal antibody specifically detects INO80E in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

INO80E
Immunofluorescence
Unconjugated
Rabbit
Human
CCDC95, coiled-coil domain containing 95, Coiled-coil domain-containing protein 95, FLJ00079, FLJ90652, INO80 complex subunit E
Produced in rabbits immunized with E. coli-derived Human INO80E fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunocytochemistry/ Immunofluorescence 1:100-1:500
Polyclonal
Purified
RUO
PBS (pH 7.0)
283899
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.