missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (2D11-H5 (same as INS05)), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP2-15195-20ug
This item is not returnable.
View return policy
Description
Insulin Monoclonal specifically detects Insulin in Human, Mouse, Porcine, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Insulin | |
| 2D11-H5 | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Insulin is a protein consisting of an alpha-chain of 21 amino acids and a beta-chain of 30 amino acids and produced in the beta-cells of the pancreas. 2D11-H5 is a specific insulin mAb as tested by ELISA and on human pancreatic tissue. Source: A BALB/c mouse was immunised with swine insulin. The mouse spleen cells were fused with SP2/0 Ag14 cells and the clones were selected on anti-insulin activity. | |
| Purified |
| Monoclonal | |
| 0.2mg/mL | |
| Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5ug/ml, SDS-Page | |
| P01308 | |
| INS | |
| The antibody reacts with the Insulin antigen. | |
| 20 ug | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| 3630 | |
| Store at 4C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction