missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (2D11-H5 (same as INS05)), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
€ 289.00 - € 504.00
Specifications
| Antigen | Insulin |
|---|---|
| Clone | 2D11-H5 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5ug/ml, SDS-Page |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18781664
|
Novus Biologicals
NBP2-15195-0.1mg |
0.1 mg |
€ 504.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18445352
|
Novus Biologicals
NBP2-15195-20ug |
20 ug |
€ 289.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Insulin Monoclonal specifically detects Insulin in Human, Mouse, Porcine, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Insulin | |
| 0.2mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| INS | |
| IgG1 | |
| Protein A or G purified | |
| Insulin is a protein consisting of an alpha-chain of 21 amino acids and a beta-chain of 30 amino acids and produced in the beta-cells of the pancreas. 2D11-H5 is a specific insulin mAb as tested by ELISA and on human pancreatic tissue. Source: A BALB/c mouse was immunised with swine insulin. The mouse spleen cells were fused with SP2/0 Ag14 cells and the clones were selected on anti-insulin activity. |
| 2D11-H5 | |
| Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5ug/ml, SDS-Page | |
| Unconjugated | |
| Mouse | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| P01308 | |
| 3630 | |
| The antibody reacts with the Insulin antigen. | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title