Learn More
Insulin R/CD220 Antibody (4B2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26192-100ul
 View more versions of this product
Product Code. 30028582
  • € 533.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human samples. It is validated for ELISA, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Insulin R/CD220
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunocytochemistry
4B2
ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR
A synthesized peptide derived from Human Insulin R/CD220 [UniProt P06213]
100 μL
Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
3643
Store at -20 to -70C. Avoid freeze-thaw cycles.