missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220 Antibody (4B2), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Insulin R/CD220 |
|---|---|
| Clone | 4B2 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30024327
|
Novus Biologicals
NBP3-26192-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028582
|
Novus Biologicals
NBP3-26192-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
| Insulin R/CD220 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
| A synthesized peptide derived from Human Insulin R/CD220 [UniProt P06213] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 4B2 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3643 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title