missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220, Rabbit anti-Human,Porcine,Bovine,Sheep, Clone: 83-14, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81038-0.025MG
This item is not returnable.
View return policy
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human, Porcine, Bovine, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation.
Specifications
| Insulin R/CD220 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognises the extracellular domain of the Insulin R/CD220. Amino acids 469 - 592 (exon 7 and 8) of Insulin R/CD220. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| 83-14 | |
| Unconjugated | |
| PBS | |
| INSR | |
| This antibody was raised by immunizing mice with IM-9 lymphocytes followed by purified insulin receptor. | |
| 0.025 mg | |
| Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3643 | |
| Human, Pig, Bovine, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction