missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220, Rabbit anti-Human,Porcine,Bovine,Sheep, Clone: 83-14, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 269.00 - € 595.00
Specifications
| Antigen | Insulin R/CD220 |
|---|---|
| Clone | 83-14 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18277598
|
Novus Biologicals
NBP2-81038-0.025MG |
0.025 mg |
€ 269.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18221909
|
Novus Biologicals
NBP2-81038-0.2MG |
0.2 mg |
€ 595.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human, Porcine, Bovine, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation.Specifications
| Insulin R/CD220 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS | |
| 3643 | |
| This antibody was raised by immunizing mice with IM-9 lymphocytes followed by purified insulin receptor. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 83-14 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Pig, Bovine, Sheep | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
| INSR | |
| IgG κ | |
| Protein A purified | |
| Recognises the extracellular domain of the Insulin R/CD220. Amino acids 469 - 592 (exon 7 and 8) of Insulin R/CD220. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title