missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22146-1-AP-150UL
This item is not returnable.
View return policy
Description
Integrin alpha 1 (ITGA1) chain associates with the beta 1 (ITGB1) chain to form a heterodimer that functions as a dual laminin/collagen receptor in neural cells and hematopoietic cells. ITGA1 has a 206-amino acid I domain in its N-terminal half, followed by 3 divalent cation-binding sites and a C-terminal transmembrane domain with a short cytoplasmic tail. It also has 28 potential N-glycosylation sites. Human ITGA1 was expressed in a mouse fibroblast cell line as a 180-kD protein. ITGA1 is involved in the early remodeling of osteoarthritic cartilage and plays an essential role in the regulation of mesenchymal stem cell proliferation and cartilage production. It also plays an essential role in the regulation of MSC proliferation and cartilage production.Specifications
| Integrin alpha-1 | |
| Polyclonal | |
| Unconjugated | |
| ITGA1 | |
| CD49a, Integrin alpha 1, integrin, alpha 1, ITGA1, Laminin and collagen receptor, VLA 1, VLA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3672 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P56199 | |
| ITGA1 | |
| Integrin alpha-1 Fusion Protein Ag17584 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction