missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | Integrin alpha-1 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16821685
|
Proteintech
22146-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16811685
|
Proteintech
22146-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha 1 (ITGA1) chain associates with the beta 1 (ITGB1) chain to form a heterodimer that functions as a dual laminin/collagen receptor in neural cells and hematopoietic cells. ITGA1 has a 206-amino acid I domain in its N-terminal half, followed by 3 divalent cation-binding sites and a C-terminal transmembrane domain with a short cytoplasmic tail. It also has 28 potential N-glycosylation sites. Human ITGA1 was expressed in a mouse fibroblast cell line as a 180-kD protein. ITGA1 is involved in the early remodeling of osteoarthritic cartilage and plays an essential role in the regulation of mesenchymal stem cell proliferation and cartilage production. It also plays an essential role in the regulation of MSC proliferation and cartilage production.Specifications
| Integrin alpha-1 | |
| Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P56199 | |
| 3672 | |
| Integrin alpha-1 Fusion Protein Ag17584 | |
| Primary | |
| -20°C | |
| ITGA1 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD49a, Integrin alpha 1, integrin, alpha 1, ITGA1, Laminin and collagen receptor, VLA 1, VLA1 | |
| ITGA1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title