Learn More
Integrin Alpha 3 Mouse, anti-Human, CoraLite Plus 488, Clone: 3F9G4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66070-100UL
 View more versions of this product
Product Code. 18018055
  • € 450.00 / 100µL
Description
The Integrin Alpha 3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Integrin Alpha 3. This antibody recognizes human, pig, mouse antigen. The Integrin Alpha 3 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
Integrin Alpha 3
3F9G4
CoraLite Plus 488
Mouse
Primary
Human, Pig, Mouse
155 kDa
IgG2a
Western Blot
1000 μg/mL
CD49C, FLJ34631, FLJ34704, FRP 2, Galactoprotein B3, GAP B3, GAPB3, Integrin alpha 3, ITGA3, MSK18, VCA 2, VL3A, VLA 3 subunit alpha, VLA3a
100 μL
3675
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.