Learn More
Integrin Alpha 3 Mouse, anti-Human, CoraLite Plus 488, Clone: 3F9G4, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | Integrin Alpha 3 |
|---|---|
| Clone | 3F9G4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18018055
|
Proteintech
CL488-66070-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18014265
|
Proteintech
CL488-66070-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Integrin Alpha 3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Integrin Alpha 3. This antibody recognizes human, pig, mouse antigen. The Integrin Alpha 3 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| Integrin Alpha 3 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 3675 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3F9G4 | |
| Western Blot | |
| Liquid | |
| Human, Pig, Mouse | |
| CD49C, FLJ34631, FLJ34704, FRP 2, Galactoprotein B3, GAP B3, GAPB3, Integrin alpha 3, ITGA3, MSK18, VCA 2, VL3A, VLA 3 subunit alpha, VLA3a | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.