missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQGAP2 Recombinant Rabbit Monoclonal Antibody (23GB4020), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553672
This item is not returnable.
View return policy
Description
Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.
Specifications
| IQGAP2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IQGAP2 | |
| 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10788 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4020 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13576 | |
| IQGAP2 | |
| A synthesized peptide derived from human IQGAP2 (1301-1495AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction