missing translation for 'onlineSavingsMsg'
Learn More

Iron phosphide, 99.5% (metals basis)

Product Code. 11365807
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365807 50 g 50g
11375807 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365807 Supplier Thermo Scientific Alfa Aesar Supplier No. 043486.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12023-53-9 | Fe3P | 198.51 g/mol

Iron phosphide is used as a semiconductor. It finds application in high power and high frequency applications like laser diodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12023-53-9
Molecular Formula Fe3P
Molecular Weight (g/mol) 198.51
MDL Number MFCD10567221
InChI Key KFSKMQQNHSLZRJ-UHFFFAOYSA-N
IUPAC Name triferriophosphane
SMILES [Fe]P([Fe])[Fe]

Specifications

Color Gray
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5% (Metals basis)
Solubility Information Soluble in nitric acid,hydrogen fluoride and aqua regia. Insoluble in water,dilute acid and alkalis.
Formula Weight 198.51
Chemical Name or Material Iron phosphide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.