missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl p-Toluenesulfonate, TRC

Product Code. 30005070
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005070 50 mg 50mg
30137672 100 mg 100mg
30136618 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005070 Supplier Toronto Research Chemicals Supplier No. TRCI77907550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4873-56-7
Molecular Formula C11 H16 O3 S
Molecular Weight (g/mol) 228.31
Synonym p-Toluenesulfonic acid isobutyl ester, 2-methylpropyl 4-methylbenzene-1-sulfonate
IUPAC Name isobutyl 4-methylbenzenesulfonate
SMILES CC(C)COS(=O)(=O)c1ccc(C)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H16O3S/c1-9(2)8-14-15(12,13)11-6-4-10(3)5-7-11/h4-7,9H,8H2,1-3H3
Formula Weight 228.082
Chemical Name or Material Isobutyl p-toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.