missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Isocitrate Dehydrogenase 2/IDH2 Antibody (SR3576), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27781-100ul
This item is not returnable.
View return policy
Description
Isocitrate Dehydrogenase 2/IDH2 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 2/IDH2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
Specifications
| Isocitrate Dehydrogenase 2/IDH2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| A synthesized peptide derived from human Isocitrate Dehydrogenase 2/IDH2 (Uniprot #: P48735) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| SR3576 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3418 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction