missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Isocitrate Dehydrogenase 2/IDH2 Antibody (SR3576), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 316.00 - € 382.00
Specifications
| Antigen | Isocitrate Dehydrogenase 2/IDH2 |
|---|---|
| Clone | SR3576 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200 |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041720
|
Novus Biologicals™
NBP3-27781-50ul |
50 μL |
€ 316.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041676
|
Novus Biologicals™
NBP3-27781-100ul |
100 μL |
€ 382.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isocitrate Dehydrogenase 2/IDH2 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 2/IDH2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western BlotSpecifications
| Isocitrate Dehydrogenase 2/IDH2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| A synthesized peptide derived from human Isocitrate Dehydrogenase 2/IDH2 (Uniprot #: P48735) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3576 | |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| 3418 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title