missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITIH5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5121815

 View more versions of this product

Product Code. 17733214

  • € 208.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody was raised against a recombinant fragment of the protein that is 25 kDa.

This gene encodes a heavy chain component of one of the inter-alpha-trypsin inhibitor (ITI) family members. ITI proteins are involved in extracellular matrix stabilization and in the prevention of tumor metastasis. They are also structurally related plasma serine protease inhibitors and are composed of a light chain and varying numbers of heavy chains. This family member is thought to function as a tumor suppressor in breast and thyroid cancers. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ITIH5
Polyclonal
Unconjugated
ITIH5
4631408O11Rik; 5430408M01Rik; DKFZp686F0145; E130106B02; inter-alpha (globulin) inhibitor H5; inter-alpha trypsin inhibitor heavy chain precursor 5; inter-alpha-inhibitor heavy chain 5; inter-alpha-trypsin inhibitor heavy chain 5; inter-alpha-trypsin inhibitor heavy chain family member 5; inter-alpha-trypsin inhibitor heavy chain family, member 5; inter-alpha-trypsin inhibitor heavy chain H5; ITI heavy chain H5; ITIH5; ITI-HC5; KIAA1953; MGC10848; PP14776; UNQ311/PRO354
Rabbit
Antigen affinity chromatography, Protein A
RUO
209378
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q8BJD1
ITIH5
Recombinant protein Inter Alpha-Globulin Inhibitor H5. The antigen corresponds to amino acid range 569-737 of the target protein.
20 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.