missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITIH5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

€ 208.00 - € 477.00

Specifications

Antigen ITIH5
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17710624
GREENER_CHOICE
View Documents
Invitrogen™
PA5119213
200 μL
€ 477.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17733214
View Documents
Invitrogen™
PA5121815
20 μL
€ 208.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody was raised against a recombinant fragment of the protein that is 25 kDa.

This gene encodes a heavy chain component of one of the inter-alpha-trypsin inhibitor (ITI) family members. ITI proteins are involved in extracellular matrix stabilization and in the prevention of tumor metastasis. They are also structurally related plasma serine protease inhibitors and are composed of a light chain and varying numbers of heavy chains. This family member is thought to function as a tumor suppressor in breast and thyroid cancers. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

ITIH5
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Mouse
ITIH5
4631408O11Rik; 5430408M01Rik; DKFZp686F0145; E130106B02; inter-alpha (globulin) inhibitor H5; inter-alpha trypsin inhibitor heavy chain precursor 5; inter-alpha-inhibitor heavy chain 5; inter-alpha-trypsin inhibitor heavy chain 5; inter-alpha-trypsin inhibitor heavy chain family member 5; inter-alpha-trypsin inhibitor heavy chain family, member 5; inter-alpha-trypsin inhibitor heavy chain H5; ITI heavy chain H5; ITIH5; ITI-HC5; KIAA1953; MGC10848; PP14776; UNQ311/PRO354
ITIH5
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q8BJD1
209378
Recombinant protein Inter Alpha-Globulin Inhibitor H5. The antigen corresponds to amino acid range 569-737 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.