missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Jagged 2 Recombinant Rabbit Monoclonal Antibody (23GB6640), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553084
This item is not returnable.
View return policy
Description
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. JAG2 is one of several ligands that activate Notch and related receptors.
Specifications
| Jagged 2 | |
| Recombinant Monoclonal | |
| 1.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97607, Q9QYE5, Q9Y219 | |
| Jag2 | |
| A synthesized peptide derived from human JAG2 (25-89AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6640 | |
| Unconjugated | |
| Jag2 | |
| D12Ggc2e; HJ2; JAG2; jagged 2; Jagged2; mJagged2-1; protein jagged-2; SER2; Serh; sm; syndactylism | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16450, 29147, 3714 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction