missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Jagged 2 Recombinant Rabbit Monoclonal Antibody (23GB6640), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | Jagged 2 |
|---|---|
| Clone | 23GB6640 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088042
|
Invitrogen™
MA553084 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089935
|
Invitrogen™
MA553085 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Jagged 2 Recombinant Monoclonal antibody specifically detects Jagged 2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Jagged 2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P97607, Q9QYE5, Q9Y219 | |
| 16450, 29147, 3714 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6640 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D12Ggc2e; HJ2; JAG2; jagged 2; Jagged2; mJagged2-1; protein jagged-2; SER2; Serh; sm; syndactylism | |
| A synthesized peptide derived from human JAG2 (25-89AA). | |
| Primary | |
| -20°C | |
| Jag2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title