missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JAK1 Recombinant Rabbit Monoclonal Antibody (23GB2245), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | JAK1 |
|---|---|
| Clone | 23GB2245 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088693
![]() |
Invitrogen™
MA553252 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088052
|
Invitrogen™
MA553253 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JAK1 Recombinant Monoclonal antibody specifically detects JAK1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| JAK1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P23458, P52332 | |
| 16451, 3716, 84598 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2245 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA960307; BAP004; C130039L05Rik; EC 2.7.10.2; Jak1; JAK-1; JAK1A; JAK1B; Janus kinase 1; Janus protein tyrosine kinase 1; JTK3; kinase Jak1; Tyrosine-protein kinase JAK1 | |
| A synthesized peptide derived from human JAK1 (625-660AA). | |
| Primary | |
| -20°C | |
| JAK1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title