missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JNJ 303

Description
Potent IKs blocker (IC50 = 64 nM). Displays no effects on other cardiac channels; IC50 values are 3.3, >10, 11.1 and 12.6 μM for INa, ICa, Ito and IKr currents respectively. Prolongs QT and causes unprovoked torsades de pointes (TdP).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 440.98 |
| Purity | >98% |
| Molecular Weight (g/mol) | 440.983 |
| Chemical Name or Material | JNJ 303 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?