missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunD Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22439
This item is not returnable.
View return policy
Description
JunD Polyclonal specifically detects JunD in Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| JunD | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| JUND | |
| Synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD. | |
| 200 μg | |
| Primary | |
| Reconstitute with 0.2 ml PBS to a final concentration of 1 mg/ml. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
| LYOPH | |
| Western Blot 10 ug/ml, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunocytochemistry/Immunofluorescence 2 - 3 ug/ml, Chromatin Immunoprecipitation (ChIP) 1-3 ul | |
| activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 3727 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction