missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunD Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 213.00 - € 519.00
Specifications
| Antigen | JunD |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 10 ug/ml, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunocytochemistry/Immunofluorescence 2 - 3 ug/ml, Chromatin Immunoprecipitation (ChIP) 1-3 ul |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ChIP Assay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18404502
|
Novus Biologicals
NBP2-22439-50ug |
50 ug |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18190472
|
Novus Biologicals
NBP2-22439 |
200 μg |
€ 519.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JunD Polyclonal specifically detects JunD in Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| JunD | |
| Western Blot 10 ug/ml, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunocytochemistry/Immunofluorescence 2 - 3 ug/ml, Chromatin Immunoprecipitation (ChIP) 1-3 ul | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 3727 | |
| Synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD. | |
| Primary | |
| Reconstitute with 0.2 ml PBS to a final concentration of 1 mg/ml. |
| LYOPH | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D | |
| JUND | |
| IgG | |
| Immunogen affinity purified | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title