missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Pop-Up Drum Fill Gauge

Prevents overfillin when dispensing waste into drums

Brand:  Justrite™ 08531

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11964957

  • € 29.50 / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

  • Features handy indicator that pops up when drum is almost full
  • Nonmetallic
  • Fits 0.75 in. (1.9cm) bung
  • Available in two lengths

Specifications

Specifications

Drum Fill Gauge
27.94 cm
11 in.
Polyethylene
To prevent overfilling when dispensing waste into drums
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers