Learn More
Invitrogen™ K-cadherin Recombinant Rabbit Monoclonal Antibody (23GB5060), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552779
 View more versions of this product
Product Code. 30089115
  • € 195.00 / 20µL
Description
Cadherins constitute a family of transmembrane glycoproteins involved in Ca2+-dependent cell-cell interactions. The members of this family are differentially expressed in various tissues. They function in the maintenance of tissue integrity and morphogenesis. Cadherins are divided into type I and type II subgroups. Type I cadherins include epithelial cadherin (E-cadherin, cadherin-1 or uvomorulin), neural cadherin (N-cadherin or cadherin-2), placental cadherin (P-cadherin or cadherin-3) and retinal cadherin (R-cadherin or cadherin-4), whereas kidney cadherin (K-cadherin or cadherin-6) and osteoblast cadherin (OB-cadherin or cadherin-11) are type II cadherins. Cadherin-6 is expressed in the proximal tubule epithelium of the kidney from which most renal cell carcinomas are derived.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
K-cadherin
Recombinant Monoclonal
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P55285, P97326
CDH6
A synthesized peptide derived from human K Cadherin (740-790AA).
20 μL
Primary
Human, Mouse
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB5060
Unconjugated
CDH6
CAD6; CADH6; cadherin 6; cadherin 6, type 2, K-cadherin; cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin6; cadherin-6; Cdh6; k cadherin; KCAD; K-cadherin; Kidney cadherin
Rabbit
Affinity chromatography
RUO
1004, 12563
-20°C