missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ K-cadherin Recombinant Rabbit Monoclonal Antibody (23GB5060), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 195.00 - € 468.00
Specifications
| Antigen | K-cadherin |
|---|---|
| Clone | 23GB5060 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087670
![]() |
Invitrogen™
MA552778 |
100 μL |
€ 468.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089115
|
Invitrogen™
MA552779 |
20 μL |
€ 195.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
K-cadherin Recombinant Monoclonal antibody specifically detects K-cadherin in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
| K-cadherin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P55285, P97326 | |
| 1004, 12563 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5060 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| CAD6; CADH6; cadherin 6; cadherin 6, type 2, K-cadherin; cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin6; cadherin-6; Cdh6; k cadherin; KCAD; K-cadherin; Kidney cadherin | |
| A synthesized peptide derived from human K Cadherin (740-790AA). | |
| Primary | |
| -20°C | |
| CDH6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title