Learn More
Ansell K200BR Heavy Duty Latex-Coated Industrial Gloves
Shop All Ansell Products
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
Description
- Seamless polycotton gloves are flexible and comfortable
- Latex-coating enhances grip capabilities in wet or dry conditions
- Palm & fully thumb coated with a crinkle finish
- Open back reduces excessive perspiration
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, REACH Compliant |
| Coating Material | Latex |
| Color | Red |
| Cut Resistance Rating | ANSI A2 |
| Exterior Finish | Fully Textured |
| Hand | Specific |
| Material | Polycotton |
| Size | 8 |
| EN388 Standard | 3241 |
| Show More |