Learn More
Poeppelmann™ Kapsto™ Polyethylene Screw Plugs with Metric Threading, GPN700 Series
Manufactured from polyethylene. Poppelmann™ Kapsto™ Polyethylene Screw Plugs with Metric Threading, GPN700 Series are suitable for a wide range of metric and imperial threads.
Brand: Poeppelmann™ 70012100000
Additional Details : Weight : 0.50000kg
Description
- Polyethylene
- Yellow
Specifications
With M12 Threading | |
Screw Plugs | |
100 Pack | |
14 mm |
Suitable for a wide range of metric and imperial threads | |
Yellow | |
Polyethylene |