Learn More
KCC2/SLC12A5-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-19565-20UL
 View more versions of this product
Product Code. 18060526
  • € 145.00 / 20µL
Description
The KCC2/SLC12A5-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human KCC2/SLC12A5-Specific. This antibody recognizes human, mouse, rat antigen. The KCC2/SLC12A5-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
KCC2/SLC12A5-Specific
1000 μg/mL
hKCC2, K Cl cotransporter 2, KCC2, KIAA1176, Neuronal K Cl cotransporter, SLC12A5
20 μL
57468
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa