Learn More
KCC2/SLC12A5-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-19565-20UL
Description
The KCC2/SLC12A5-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human KCC2/SLC12A5-Specific. This antibody recognizes human, mouse, rat antigen. The KCC2/SLC12A5-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| KCC2/SLC12A5-Specific | |
| 1000 μg/mL | |
| hKCC2, K Cl cotransporter 2, KCC2, KIAA1176, Neuronal K Cl cotransporter, SLC12A5 | |
| 20 μL | |
| 57468 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |