Learn More
KCC2/SLC12A5-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | KCC2/SLC12A5-Specific |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18060526
|
Proteintech
CL488-19565-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18016215
|
Proteintech
CL488-19565-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The KCC2/SLC12A5-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human KCC2/SLC12A5-Specific. This antibody recognizes human, mouse, rat antigen. The KCC2/SLC12A5-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| KCC2/SLC12A5-Specific | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| hKCC2, K Cl cotransporter 2, KCC2, KIAA1176, Neuronal K Cl cotransporter, SLC12A5 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 57468 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.