Learn More
KIAA1755 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-06014-100ul
 View more versions of this product
Product Code. 18609228
  • € 326.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
KIAA1755 Polyclonal antibody specifically detects KIAA1755 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
KIAA1755
Polyclonal
Immunohistochemistry-Paraffin 1:50-1:200
KIAA1755 KIAA1755, RP5-1054A22.3
Produced in rabbits immunized with E. coli-derived Human KIAA1755 fragment.
100 μg
Primary
Human
Purified
Immunohistochemistry (Paraffin)
Unconjugated
PBS, pH7.0
Rabbit
Antigen and protein A Affinity-purified
RUO
85449
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.