missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1755 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 231.00 - € 326.00
Specifications
| Antigen | KIAA1755 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18609228
|
Novus Biologicals
NBP3-06014-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660578
|
Novus Biologicals
NBP3-06014-50ul |
50 μg |
€ 231.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIAA1755 Polyclonal antibody specifically detects KIAA1755 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| KIAA1755 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| KIAA1755 KIAA1755, RP5-1054A22.3 | |
| Produced in rabbits immunized with E. coli-derived Human KIAA1755 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 85449 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title