missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kif4A Antibody (SR1195), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22228-100ul
This item is not returnable.
View return policy
Description
Kif4A Monoclonal antibody specifically detects Kif4A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Kif4A | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR1195 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100 | |
| chromokinesin-A, chromosome-associated kinesin KIF4A, FLJ12530, FLJ12655, FLJ14204, FLJ20631, HSA271784, KIF4G1, KIF4KIF4-G1, kinesin family member 4A | |
| A synthesized peptide derived from human Kif4A (Uniprot #: O95239) | |
| 100 μg | |
| Core ESC Like Genes, Lipid and Metabolism, Mitotic Regulators, Neuroscience, Stem Cell Markers | |
| 24137 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction