missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kif4A Antibody (SR1195), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Kif4A |
|---|---|
| Clone | SR1195 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671765
|
Novus Biologicals
NBP3-22228-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692275
|
Novus Biologicals
NBP3-22228-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kif4A Monoclonal antibody specifically detects Kif4A in Human, Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
| Kif4A | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| chromokinesin-A, chromosome-associated kinesin KIF4A, FLJ12530, FLJ12655, FLJ14204, FLJ20631, HSA271784, KIF4G1, KIF4KIF4-G1, kinesin family member 4A | |
| A synthesized peptide derived from human Kif4A (Uniprot #: O95239) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1195 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Lipid and Metabolism, Mitotic Regulators, Neuroscience, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 24137 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title