Learn More
DWK Life Sciences Kimble™ Aluminum Seals for Headspace Vials
Allow for easy access to vial contents
Brand: DWK Life Sciences 73820-20
Additional Details : Weight : 0.10000kg
Description
Allow for easy access to vial contents
- One piece
- Fits GPI Aluminum Seal Finish 20/20A
- Unlined
- Aluminum
- 20mm (0.787 in.) size
Model 73820-20
- Tear-out style
- Tear-off style
Specifications
Natural | |
1000 Cs. | |
20 mm |
Aluminum | |
Tear-Out Crimp |