Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials with Attached White Rubber Lined Closures
Attached closed top black phenolic closures
€ 203.00 - € 347.00
Specifications
Material (Liner) | Rubber |
---|---|
Color | Amber |
Material | Borosilicate Glass |
Thread Style | Screw |
Closure Material | Phenolic |
Product Code | Brand | Closure Type | Volume (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Volume (English) | Price | Quantity & Availability | ||||
11843023
|
DWK Life Sciences
60911D12 |
Screw Cap | 0.37 dr. |
€ 347.00
Pack of 288 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
||||
11872973
|
DWK Life Sciences
60911D1 |
Screw Cap (Closed-top) | 1 dr. |
€ 203.00
Pack of 144 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
||||
11882973
|
DWK Life Sciences
60911D2 |
Screw Cap (Closed-top) | 2.16 dr. |
€ 223.00
Pack of 144 |
Estimated Shipment: 01-07-2024
Log in to see stock. |
||||
Description
Attached closed top black phenolic closures
- Manufactured from amber 51 expansion borosilicate glass
- Closures with white rubber liners
- Vials packaged in corrugated trays with partitions
Specifications
Rubber | |
Borosilicate Glass | |
Phenolic |
Amber | |
Screw | |
USP Type I |