Learn More
DWK Life Sciences Kimble™ KIMAX™ Accuform™ Micro-Vial™ Vials
Product Code. 10197908
Click to view available options
Volume (Metric):
0.1 mL
0.3 mL
1.0 mL
5.0 mL
10.0 mL
Color:
Clear
Graduated:
No
Yes
Volume (English):
0.03 dr.
0.08 dr.
0.25 dr.
1.35 dr.
2.75 dr.
Unit Size:
Pack of 12
Product Code. 10197908
Brand: DWK Life Sciences 607101
Only null left
Add to basket
Add to basket
Conical cavity allows maximum retrieval of contents with a syringe needle.
  • Clear glass Micro-Vial with open top closure and PTFE-faced silicone septum
  • Made with extra thick clear glass walls for both sturdiness and magnification of contents
  • Ungraduated, with marking spot
  • Tube has a screw thread finish
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Silicone
Color Clear
Diameter (Metric) Outer 14 mm
Material Borosilicate Glass
Volume (Metric) 1.0 mL
Graduated No
Style Tapered
Thread Style Screw
Volume (English) 0.25 dr.
Closure Type Screw Cap (Open-top)
Height (Metric) 49 mm
Certifications/Compliance ASTM E438, USP Type I, Type I, Class A