Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Bonded White PTFE Liners
Click to view available options
Color:
Black
Blue
White
Closure Size:
13-425
8-425
9-425
Unit Size:
Pack of 1000
Description
- Manufactured using the highest quality raw materials
- Designed for an ideal fit with Kimble Chase autosampler vials
- Pre-assembled caps and septa minimizes contamination due to handling
Specifications
Specifications
| Material (Liner) | PTFE |
| Color | Black |
| Material | Polypropylene |
| Autoclavable | Autoclavable |
| Closure Size | 13-425 |