Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached White Rubber Lined Caps
Product Code. 11842973
Click to view available options
Volume (Metric):
2 mL
4 mL
6 mL
8 mL
12 mL
16 mL
20 mL
24 mL
40 mL
Height (Metric):
28 mm
35 mm
38 mm
40 mm
45 mm
57 mm
60 mm
65 mm
70 mm
85 mm
95 mm
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
21 mm
23 mm
28 mm
Quantity:
144/Cs.
200/Cs.
288/Cs.
72/Cs.
Unit Size:
Pack of 144
Pack of 200
Pack of 288
Pack of 72
Product Code. 11842973
Brand: DWK Life Sciences 60811D6
Only null left
Add to basket
Add to basket
Packaged in corrugated trays with partitions
  • Small case quantities
  • Manufactured from clear 33 expansion borosilicate glass
  • Attached closed top black phenolic closures
TRUSTED_SUSTAINABILITY
Specifications
Classification A
Material (Liner) Rubber
Color Clear
Diameter (Metric) Outer 23 mm
Material Borosilicate Glass
Volume (Metric) 24 mL
Thread Style Screw
Volume (English) 6.5 dr.
Closure Material Phenolic
Closure Type Screw Cap (Closed-top)
Height (Metric) 85 mm
Quantity 144/Cs.