Learn More
HMC Europe Vertical Autoclave Air Filters
Exhaust air filter. Safety for individuals and the environment. Option for safe, emission-free sterilization of highly infectious germs and genetically modified materials. Not for applications requiring FDA approval.
Brand: HMC Europe 50004
Additional Details : Weight : 0.50000kg
Description
During the venting phase, the steam displaces unsterilized residual air from the chamber and passes it through a filter system. The surrounding air is protected against contamination as the cycle is running. Naturally, in-line sterilization of the exhaust air filter is performed. Due to the design, condensate sterilization is a matter of course. Any condensate which precipitates flows back into the chamber and gets sterilized. Discharge can be done safely.Specifications
Sterilization of exhaust air | |
HMC-HV autoclave | |
Exhaust Air Filter |