missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLC2 Monoclonal Antibody (OTI10H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811478
This item is not returnable.
View return policy
Description
Kinesin is a molecular motor that generates ATP-dependent movement of vesicles and organelles along microtubules. Kinesin consists of 2 light chains, such as KLC2, and 2 heavy chains in a 1:1 stoichiometric ratio.Specifications
| KLC2 | |
| Monoclonal | |
| Unconjugated | |
| KLC2 | |
| kinesin light chain 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64837 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10H10 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H0B6 | |
| KLC2 | |
| Human recombit protein fragment corresponding to amino acids 335-622 of human KLC2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction