Learn More
Invitrogen™ KLC3 Recombinant Rabbit Monoclonal Antibody (24GB3760)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557047
This item is not returnable.
View return policy
Description
KLC3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
KLC3 encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria.
Specifications
| KLC3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| KLC3 | |
| BC017147; kinesin light chain 2; kinesin light chain 3; Kinesin light chain KLCt; KLC2; KLC2L; KLC2-like; Klc3; Klct; KNS2B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 147700, 171549, 232943 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3760 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q68G30, Q6P597, Q91W40 | |
| KLC3 | |
| A synthesized peptide derived from human KLC3 (aa 350-410). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |