missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KLC3 Recombinant Rabbit Monoclonal Antibody (24GB3760)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

€ 146.00 - € 500.00

Specifications

Antigen KLC3
Clone 24GB3760
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276091
GREENER_CHOICE
View Documents
Invitrogen™
MA557047
100 μL
€ 500.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276656
View Documents
Invitrogen™
MA557048
20 μL
€ 146.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KLC3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

KLC3 encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria.
Specifications

Specifications

KLC3
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
KLC3
BC017147; kinesin light chain 2; kinesin light chain 3; Kinesin light chain KLCt; KLC2; KLC2L; KLC2-like; Klc3; Klct; KNS2B
KLC3
Primary
Affinity chromatography
24GB3760
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q68G30, Q6P597, Q91W40
147700, 171549, 232943
A synthesized peptide derived from human KLC3 (aa 350-410).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.