Learn More
Invitrogen™ KLC3 Recombinant Rabbit Monoclonal Antibody (24GB3760)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557048
 View more versions of this product
Product Code. 30276656
  • € 146.00 / 20µL
Description
KLC3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
KLC3 encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KLC3
Recombinant Monoclonal
Unconjugated
KLC3
BC017147; kinesin light chain 2; kinesin light chain 3; Kinesin light chain KLCt; KLC2; KLC2L; KLC2-like; Klc3; Klct; KNS2B
Rabbit
Affinity chromatography
RUO
147700, 171549, 232943
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3760
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q68G30, Q6P597, Q91W40
KLC3
A synthesized peptide derived from human KLC3 (aa 350-410).