missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLH Antibody (15F7E4G3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05779-100ul
This item is not returnable.
View return policy
Description
KLH Monoclonal antibody specifically detects KLH in Nonspecies specific samples. It is validated for Western Blot, ELISA
Specifications
| KLH | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Western Blot, ELISA | |
| 15F7E4G3 | |
| Western Blot 1:500-1:1000, ELISA 1:1000-1:2000 | |
| keyhole limpet hemocyanin (KLH) | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant KLH | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction