missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLH Antibody (15F7E4G3), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 302.00
Specifications
| Antigen | KLH |
|---|---|
| Clone | 15F7E4G3 |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:1000-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603219
|
Novus Biologicals
NBP3-05779-100ul |
100 μg |
€ 302.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635896
|
Novus Biologicals
NBP3-05779-50ul |
50 μg |
€ 220.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLH Monoclonal antibody specifically detects KLH in Nonspecies specific samples. It is validated for Western Blot, ELISASpecifications
| KLH | |
| Western Blot 1:500-1:1000, ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant KLH | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 15F7E4G3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| keyhole limpet hemocyanin (KLH) | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title